ADVH49B82949 AChemBlock

tert-butyl 3-[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]azetidine-1-carboxylate

  • Molecular Weight : 349.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84657
Availability details for
Quantity
  • Availability
  • Add to Cart