ADVH49B8294C AChemBlock

tert-butyl 5-(hydroxymethyl)-octahydrocyclopenta[b]pyrrole-1-carboxylate

  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-hydroxymethyl-octahydrocyclopenta-b-pyrrole-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84609
Availability details for
Quantity
  • Availability
  • Add to Cart