ADVH49B8294F AChemBlock

tert-butyl (4S)-4-amino-3,3-difluoropiperidine-1-carboxylate

  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4s-4-amino-3-3-difluoropiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84734
Availability details for
Quantity
  • Availability
  • Add to Cart