ADVH49B82950 AChemBlock

ethyl 5-methanesulfonyl-2,3-dihydro-1H-indole-3-carboxylate

  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-methanesulfonyl-2-3-dihydro-1h-indole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85024
Availability details for
Quantity
  • Availability
  • Add to Cart