ADVH49B82953 AChemBlock

ethyl 5,5-difluoro-1,4,6,7-tetrahydroindazole-3-carboxylate

  • Molecular Weight : 230.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-5-difluoro-1-4-6-7-tetrahydroindazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85027
Availability details for
Quantity
  • Availability
  • Add to Cart