ADVH49B82955 AChemBlock

ethyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-3-carboxylate

  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85029
Availability details for
Quantity
  • Availability
  • Add to Cart