ADVH49B82957 AChemBlock

ethyl 3-fluoroimidazo[1,2-a]pyridine-2-carboxylate

  • Molecular Weight : 208.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-fluoroimidazo-1-2-a-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85031
Availability details for
Quantity
  • Availability
  • Add to Cart