ADVH49B82959 AChemBlock

ethyl 3-amino-4-hydroxy-pyridine-2-carboxylate

  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-amino-4-hydroxy-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85033
Availability details for
Quantity
  • Availability
  • Add to Cart