ADVH49B8295E AChemBlock

tert-butyl (4R)-4-amino-2,2-difluoro-8-azaspiro[4.5]decane-8-carboxylate

  • Molecular Weight : 290.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4r-4-amino-2-2-difluoro-8-azaspiro-4-5-decane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84741
Availability details for
Quantity
  • Availability
  • Add to Cart