ADVH49B8295F AChemBlock

ethyl 2-[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]acetate

  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrimidin-2-yl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85038
Availability details for
Quantity
  • Availability
  • Add to Cart