ADVH49B82960 AChemBlock

tert-butyl 3,6-diazabicyclo[3.2.0]heptane-3-carboxylate;hydrochloride

  • Molecular Weight : 234.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-6-diazabicyclo-3-2-0-heptane-3-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84660
Availability details for
Quantity
  • Availability
  • Add to Cart