ADVH49B82962 AChemBlock

ethyl 2-[(thietan-3-yl)amino]propanoate

  • Molecular Weight : 189.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-thietan-3-yl-amino-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85041
Availability details for
Quantity
  • Availability
  • Add to Cart