ADVH49B82964 AChemBlock

ethyl 2-(4-aminocyclohexyl)propanoate

  • Molecular Weight : 199.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-4-aminocyclohexyl-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85043
Availability details for
Quantity
  • Availability
  • Add to Cart