ADVH49B82968 AChemBlock

tert-butyl 3,3-difluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate

  • Molecular Weight : 347.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-3-difluoro-4-tetramethyl-1-3-2-dioxaborolan-2-yl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84663
Availability details for
Quantity
  • Availability
  • Add to Cart