ADVH49B82969 AChemBlock

ethyl (3S,4R)-4-[tert-butoxycarbonyl(methyl)amino]pyrrolidine-3-carboxylate

  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3s-4r-4-tert-butoxycarbonyl-methyl-amino-pyrrolidine-3-carboxylate-308358.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85048
Availability details for
Quantity
  • Availability
  • Add to Cart