ADVH49B8296B AChemBlock

tert-butyl (3S,4R)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(hydroxymethyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 438.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-4r-3-9h-fluoren-9-yl-methoxy-carbonyl-amino-4-hydroxymethyl-pyrrolidine-1-carboxylate-308056.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84746
Availability details for
Quantity
  • Availability
  • Add to Cart