ADVH49B8296F AChemBlock

ethyl (2R,5S)-5-(benzyloxyamino)piperidine-2-carboxylate

  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2r-5s-5-benzyloxyamino-piperidine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85054
Availability details for
Quantity
  • Availability
  • Add to Cart