ADVH49B82972 AChemBlock

tert-butyl (3S)-3-amino-1-oxa-6-azaspiro[3.3]heptane-6-carboxylate

  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-amino-1-oxa-6-azaspiro-3-3-heptane-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84748
Availability details for
Quantity
  • Availability
  • Add to Cart