ADVH49B82973 AChemBlock

tert-butyl (3S)-3-{[(tert-butoxy)carbonyl]amino}-3-(hydroxymethyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 316.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-tert-butoxy-carbonyl-amino-3-hydroxymethyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84749
Availability details for
Quantity
  • Availability
  • Add to Cart