ADVH49B82974 AChemBlock

tert-butyl (3S)-3-[tert-butyl(dimethyl)silyl]oxy-5-oxo-piperidine-1-carboxylate

  • Molecular Weight : 329.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-tert-butyl-dimethyl-silyl-oxy-5-oxo-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84750
Availability details for
Quantity
  • Availability
  • Add to Cart