ADVH49B82977 AChemBlock

tert-butyl (3S)-3-(aminomethyl)-3-methyl-piperidine-1-carboxylate

  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-aminomethyl-3-methyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84751
Availability details for
Quantity
  • Availability
  • Add to Cart