ADVH49B8297D AChemBlock

dimethyl[(2R)-pyrrolidin-2-ylmethyl]amine dihydrochloride

  • Molecular Weight : 201.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-2r-pyrrolidin-2-ylmethyl-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85068
Availability details for
Quantity
  • Availability
  • Add to Cart