ADVH49B8297E AChemBlock

dimethyl (1r,4r)-bicyclo[2.1.1]hexane-1,4-dicarboxylate

  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-1r-4r-bicyclo-2-1-1-hexane-1-4-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71898
Availability details for
Quantity
  • Availability
  • Add to Cart