ADVH49B82980 AChemBlock

cis-octahydropyrano[3,4-c]pyrrole

  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-octahydropyrano-3-4-c-pyrrole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85070
Availability details for
Quantity
  • Availability
  • Add to Cart