ADVH49B82983 AChemBlock

cis-N,N,2-trimethylpyrrolidin-3-amine

  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-n-n-2-trimethylpyrrolidin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85073
Availability details for
Quantity
  • Availability
  • Add to Cart