ADVH49B82984 AChemBlock

cis-hexahydro-1H-furo[3,4-c]pyrrol-4-one

  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-hexahydro-1h-furo-3-4-c-pyrrol-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85074
Availability details for
Quantity
  • Availability
  • Add to Cart