ADVH49B82986 AChemBlock

cis-8-methyl-3,8-diazabicyclo[4.2.0]octane hydrochloride

  • Molecular Weight : 162.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-8-methyl-3-8-diazabicyclo-4-2-0-octane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85076
Availability details for
Quantity
  • Availability
  • Add to Cart