ADVH49B82988 AChemBlock

cis-5-Aminotetrahydro-2H-pyran-3-ol hydrochloride

  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-5-aminotetrahydro-2h-pyran-3-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85078
Availability details for
Quantity
  • Availability
  • Add to Cart