ADVH49B82989 AChemBlock

cis-5-{[(tert-butoxy)carbonyl]amino}oxane-3-carboxylic acid

  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-5-tert-butoxy-carbonyl-amino-oxane-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85079
Availability details for
Quantity
  • Availability
  • Add to Cart