ADVH49B8298B AChemBlock

tert-butyl (3S)-3-(2-morpholinoethyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 284.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-2-morpholinoethyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84752
Availability details for
Quantity
  • Availability
  • Add to Cart