ADVH49B8298E AChemBlock

tert-butyl (3R,4S)-rel-3-(hydroxymethyl)-4-methoxypyrrolidine-1-carboxylate

  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-4s-rel-3-hydroxymethyl-4-methoxypyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84754
Availability details for
Quantity
  • Availability
  • Add to Cart