ADVH49B82990 AChemBlock

cis-3-fluoro-4-(methoxymethyl)piperidine hydrochloride

  • Molecular Weight : 183.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-3-fluoro-4-methoxymethyl-piperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85086
Availability details for
Quantity
  • Availability
  • Add to Cart