ADVH49B82995 AChemBlock

cis-2-methoxycyclopropanamine hydrochloride

  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-2-methoxycyclopropanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85091
Availability details for
Quantity
  • Availability
  • Add to Cart