ADVH49B82998 AChemBlock

cis-2-aminocyclopentane-1-carbonitrile hydrochloride

  • Molecular Weight : 146.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-2-aminocyclopentane-1-carbonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85094
Availability details for
Quantity
  • Availability
  • Add to Cart