ADVH49B8299A AChemBlock

cis-2-(tert-butoxycarbonylamino)-5,5-difluoro-cyclohexanecarboxylic acid

  • Molecular Weight : 279.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-2-tert-butoxycarbonylamino-5-5-difluoro-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85096
Availability details for
Quantity
  • Availability
  • Add to Cart