ADVH49B8299D AChemBlock

cis-1-[(tert-butoxy)carbonyl]-4-methylpyrrolidine-3-carboxylic acid

  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-1-tert-butoxy-carbonyl-4-methylpyrrolidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85099
Availability details for
Quantity
  • Availability
  • Add to Cart