ADVH49B829A0 AChemBlock

tert-butyl (3R,4S)-3-(hydroxymethyl)-4-(2-methylpropyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 257.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-4s-3-hydroxymethyl-4-2-methylpropyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84756
Availability details for
Quantity
  • Availability
  • Add to Cart