ADVH49B829A2 AChemBlock

CDDO-Im

  • Molecular Weight : 541.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.achemblock.com/cddo-im.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86463
Availability details for
Quantity
  • Availability
  • Add to Cart