ADVH49B829A4 AChemBlock

tert-butyl (3R,4R)-3-methyl-4-(methylamino)pyrrolidine-1-carboxylate

  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-4r-3-methyl-4-methylamino-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84758
Availability details for
Quantity
  • Availability
  • Add to Cart