ADVH49B829A5 AChemBlock

tert-butyl (3R)-4,4-difluoro-3-hydroxypiperidine-1-carboxylate

  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-4-4-difluoro-3-hydroxypiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84759
Availability details for
Quantity
  • Availability
  • Add to Cart