ADVH49B829A7 AChemBlock

benzyl N-{1-oxa-7-azaspiro[3.5]nonan-3-yl}carbamate

  • Molecular Weight : 276.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-1-oxa-7-azaspiro-3-5-nonan-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85109
Availability details for
Quantity
  • Availability
  • Add to Cart