ADVH49B829AA AChemBlock

benzyl N-{[(3R,4S)-3-fluoropiperidin-4-yl]methyl}carbamate

  • Molecular Weight : 266.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-3r-4s-3-fluoropiperidin-4-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85112
Availability details for
Quantity
  • Availability
  • Add to Cart