ADVH49B829AD AChemBlock

benzyl N-[(3S,5R)-5-methylpiperidin-3-yl]carbamate

  • Molecular Weight : 248.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-3s-5r-5-methylpiperidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85115
Availability details for
Quantity
  • Availability
  • Add to Cart