ADVH49B829AE AChemBlock

benzyl N-[(3S,4S)-4-methoxypyrrolidin-3-yl]-N-methylcarbamate

  • Molecular Weight : 264.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-3s-4s-4-methoxypyrrolidin-3-yl-n-methylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85116
Availability details for
Quantity
  • Availability
  • Add to Cart