ADVH49B829AF AChemBlock

benzyl N-[(1S)-4-oxocyclopent-2-en-1-yl]carbamate

  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-1s-4-oxocyclopent-2-en-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85117
Availability details for
Quantity
  • Availability
  • Add to Cart