ADVH49B829B0 AChemBlock

benzyl N-[(1R,2R,3R,5R)-2-[tert-butyl(dimethyl)silyl]oxy-3-bicyclo[3.1.0]hexanyl]carbamate

  • Molecular Weight : 361.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-1r-2r-3r-5r-2-tert-butyl-dimethyl-silyl-oxy-3-bicyclo-3-1-0-hexanyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85118
Availability details for
Quantity
  • Availability
  • Add to Cart