ADVH49B829B3 AChemBlock

benzyl N-({1-oxa-7-azaspiro[3.5]nonan-2-yl}methyl)carbamate

  • Molecular Weight : 290.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-n-1-oxa-7-azaspiro-3-5-nonan-2-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85121
Availability details for
Quantity
  • Availability
  • Add to Cart