ADVH49B829B8 AChemBlock

benzyl cis-3-(aminomethyl)-2-methylpiperidine-1-carboxylate

  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-cis-3-aminomethyl-2-methylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85126
Availability details for
Quantity
  • Availability
  • Add to Cart