ADVH49B829BA AChemBlock

tert-butyl 3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-5H,6H,7H-pyrrolo[3,4-b]pyridine-6-carboxylate

  • Molecular Weight : 346.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-tetramethyl-1-3-2-dioxaborolan-2-yl-5h-6h-7h-pyrrolo-3-4-b-pyridine-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84665
Availability details for
Quantity
  • Availability
  • Add to Cart