ADVH49B829BC AChemBlock

benzyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindoline-2-carboxylate

  • Molecular Weight : 379.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-isoindoline-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85130
Availability details for
Quantity
  • Availability
  • Add to Cart